Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Product Code. p-4526364
Change view
Click to view available options
Marking Spot:
No
Yes
Closure Color:
Black
Light Blue
Septum:
PTFE/Silicone, Pre-slit
Red PTFE/White Silicone
Color:
Amber
Clear
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Marking Spot Closure Color Septum Color unitSize
12463608 No Light Blue PTFE/Silicone, Pre-slit Clear Pack of 100
12433628 No Light Blue Red PTFE/White Silicone Amber Pack of 100
12453628 Yes Light Blue Red PTFE/White Silicone Amber Pack of 100
12463628 No Black Red PTFE/White Silicone Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12463608 Supplier Thermo Scientific™ Supplier No. C4010018
Only null left
Add to basket
Add to basket
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
  • Assembled kits include 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers or in polybags
  • Vial trays are stackable for easy storage
  • Vial trays hold vials upright for easy filling and maintaining sample order
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Light Blue
Description Clear Glass; Light Blue Cap; PTFE/Silicone Pre-slit septum
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Pre-slit Yes
Septum PTFE/Silicone, Pre-slit
Marking Spot No