missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Assembled Convenience Kits: EPA Vials with Caps and Septa

Product Code. 12498646
Change view
Click to view available options
Volume (Metric):
40 mL
Color:
Clear
Certifications/Compliance:
Class 200
Class 300
Unit Size:
Pack of 72
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Certifications/Compliance unitSize
12498646 40 mL Clear Class 200 Pack of 72
12408656 40 mL Clear Class 300 Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12498646 Supplier Thermo Scientific™ Supplier No. B7950C

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.

  • Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water”
  • Class 100—Vials, Caps and Septa are assembled
  • Class 200—Vials, Caps and Septa are processed to EPA specifications and assembled
  • Class 300—Vials, Caps and Septa are processed and certified to EPA specifications, then assembled

Includes:

Certificate of Analysis is included with each box of vials.

TRUSTED_SUSTAINABILITY

Specifications

Description Clear; 40mL
Color Clear
Volume (Metric) 40 mL
Closure Size 24-400
For Use With (Equipment) Discrete Water Sampling Applications
Certifications/Compliance Class 200
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.