Learn More
Naspm trihydrochloride, MedChemExpress
Product Code. 16461437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16461437
Brand: Medchem Express HY12506A
MedChemExpress Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Specifications
Chemical Name or Material Naspm trihydrochloride
Physical Form Solid
CAS 1049731-36-3
Color Pale Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H37Cl3N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym 1-Naphthylacetyl spermine trihydrochloride
Solubility Information H2O : 50 mg/mL (104.19 mM; Need ultrasonic) ∣DMSO : 6.4 mg/mL (13.34 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.[H]Cl.O=C(NCCCNCCCCNCCCN)CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 479.91
Formula Weight 479.91
Percent Purity 98.0%
Grade Research