missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nardilysin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93058-0.02ml
This item is not returnable.
View return policy
Description
Nardilysin Polyclonal antibody specifically detects Nardilysin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Nardilysin | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| EC 3.4.24, EC 3.4.24.61, hNRD1, hNRD2, nardilysin, nardilysin (N-arginine dibasic convertase), N-arginine dibasic convertase, NRD convertase, NRD-C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-400 of human NRD1 (NP_001095132.1). SGAEIEDDDEEGFDDEDEFDDEHDDDLDTEDNELEELEERAEARKKTTEKQSAAALCVGVGSFADPDDLPGLAHFLEHMVFMGSLKYPDENGFDAFLKKHGGSDNASTDCERTVFQFDVQRKYFKEALDRWAQFFIHPLMIRDAIDREVEAVDSEYQLARPSDANRKEMLFGSLARPGHPMGKFFWGNAETLKHEPRKNNIDTHARLREFWMRYYSSHYMTLVVQSKETLDTLEKWVTEIF | |
| 0.02 mL | |
| Neuroscience | |
| 4898 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction