Learn More
NaPi-IIa Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26783-1-AP-20UL
Description
The NaPi-IIa antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NaPi-IIa. This antibody recognizes human, mouse, rat antigen. The NaPi-IIa antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NaPi-IIa | |
| 750 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| Na(+)/Pi cotransporter 2A, NaPi 2a, NAPI 3, napi2a, NaPi-IIa, SLC11, SLC17A2, SLC34A1 | |
| 20 μL | |
| 6569 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.