Learn More
NaPi-IIa Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26783-1-AP-150UL
 View more versions of this product
Product Code. 18076395
  • 410.00 EUR / 150µL
Description
The NaPi-IIa antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NaPi-IIa. This antibody recognizes human, mouse, rat antigen. The NaPi-IIa antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NaPi-IIa
750 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
Na(+)/Pi cotransporter 2A, NaPi 2a, NAPI 3, napi2a, NaPi-IIa, SLC11, SLC17A2, SLC34A1
150 μL
6569
Store at -20°C. Stable for one year after shipment.
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions