missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 35245-26-2 |
| Color | White |
| Molecular Formula | C20H16ClNO3 |
| Quantity | 1 g |
| Synonym | naphthol as-d chloroacetate, 2-o-tolylcarbamoyl naphthalen-3-yl 2-chloroacetate, 3-hydroxy-2-naphthoic-o-toluidide chloroacetate, naphthol as-d-chloroacetate, 3-2-methylphenyl carbamoyl-2-naphthyl chloroacetate, 3-2-methylphenyl carbamoyl naphthalen-2-yl 2-chloroacetate, acetic acid, 2-chloro-,3-2-methylphenyl amino carbonyl-2-naphthalenyl ester, naphthol-as-d-chloroacetate, acetic acid, chloro-, s, naphthol as-d chloroacetate solution |
| InChI Key | FMVKYSCWHDVMGO-UHFFFAOYSA-N |
| SMILES | CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2OC(=O)CCl |
| IUPAC Name | [3-[(2-methylphenyl)carbamoyl]naphthalen-2-yl] 2-chloroacetate |
| Molecular Weight (g/mol) | 353.802 |
| PubChem CID | 160247 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?