Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Color | Gray |
| Description | EFA: 0.3 cm2 |
| Material (Membrane) | PES |
| MWCO | 3 kDa |
| Sterility | Non-sterile |
| Housing Material | Polypropylene |
| Includes | 100/Pk. |
| Volume (Metric) | 50 to 500 μL (Sample) |