Learn More
Thermo Scientific NanoDrop™ Reagents
Shop All Thermo Scientific Products
Click to view available options
Type:
PV-1 Performance Verification
PV-8 Performance Verification
Unit Size:
Each
Description
Certified standards for verifying instrument performance to factory specifications are available separately.
Features include:
- Solution with certified absorbance values
- Use to create a record of correct photometric performance of a NanoDrop One spectrophotometer
Specifications
Specifications
| Type | PV-1 Performance Verification |
| For Use With (Equipment) | NanoDrop Ultra, NanoDrop One and NanoDrop OneC spectrophotometers |
| Storage Requirements | Store at room temperature |