missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Acetyl-D-arginine dihydrate, 98%

Product Code. p-7024235
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11335209

Brand: Thermo Scientific Alfa Aesar A18563.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 210545-23-6 | C8H16N4O3 | 216.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210545-23-6
Molecular Formula C8H16N4O3
Molecular Weight (g/mol) 216.24
MDL Number MFCD00209627
InChI Key SNEIUMQYRCDYCH-ZCFIWIBFSA-N
Synonym ac-d-arg-oh, nalpha-acetyl-d-arginine dihydrate, n-alpha-acetyl-d-arginine dihydrate, 2r-5-carbamimidamido-2-acetamidopentanoic acid dihydrate, n-?-acetyl-d-arginine dihydrate, n-, a-acetyl-d-arginine dihydrate, 2r-5-diaminomethylidene amino-2-acetamidopentanoic acid dihydrate
PubChem CID 72698573
IUPAC Name (2R)-5-{[amino(iminiumyl)methyl]amino}-2-acetamidopentanoate
SMILES CC(=O)N[C@H](CCCNC(N)=[NH2+])C([O-])=O

Specifications

Melting Point ∼270°C
Quantity 1 g
Formula Weight 252.29 (216.25 Anhydrous)
Percent Purity 99%
Chemical Name or Material Nalpha-Acetyl-D-arginine dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.