Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
GREENER_CHOICE
Product Code. p-3341498
Click to view available options
For Use With (Equipment):
Replacement O-ring (for Mfr. No. DS3132-0058)
Replacement O-ring (for Mfr. No. DS3132-0063)
Unit Size:
Pack of 10
Product Code. 10420533
Brand: Thermo Scientific™ 7131320063
This item requires a price update prior to being ordered.
Request a Price for this item here.
Ensure leakproof service at their maximum speeds >10,000 x g, even when full
  • Polypropylene closures with silicone gasket
Before autoclaving, set closure on top of the container without engaging the threads.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Silicone
Tip Material Silicone
For Use With (Equipment) Replacement O-ring (for Mfr. No. DS3132-0063)