Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Shop All Thermo Scientific Products
Click to view available options
Holds:
1 and 1.5 mL Vials
2 mL Vials
5 mL Vials
Vial Capacity:
25
81
100
Quantity:
1/Cs.
10/Cs.
20/Cs.
40/Cs.
80/Cs.
Unit Size:
Pack of 10
Pack of 20
Pack of 25
Pack of 40
Pack of 81
Description
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
- Warranty varies by country, please contact us for details
Specifications
Specifications
| Type | Cryogenic Storage Box |
| Capacity (English) | 7.06 cu. ft. |
| Capacity (Metric) | 2 mL |
| For Use With (Equipment) | Locator 8 Plus |
| Height (English) Exterior | 2 in. |
| Vial Capacity | 25 |
| Holds | 2 mL Vials |
| Quantity | 80/Cs. |