Learn More
Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels
GREENER_CHOICE
Product Code. p-4525022
Click to view available options
Capacity (Metric):
83 mL
186 mL
358 mL
919 mL
2.91 L
Diameter (Metric)Top:
58 mm
75 mm
99 mm
132 mm
175 mm
Unit Size:
Each
Product Code. 10657711
Brand: Thermo Scientific™ 42800550
Only null left
Add to basket
Add to basket
Separate the lightweight and durable two-piece polypropylene funnels for easy cleaning. These funnels won't chip, crack or break.
Separate the lightweight and durable Thermo Scientific™ Nalgene™ Büchner Two-Piece Polypropylene Funnels for easy cleaning. These autoclavable funnels won't chip, crack or break making them a valuble asset to lab safety programs.
  • Sturdy funnels consist of cup and funnel with perforated plate for easy cleaning and good chemical resistance
  • All sizes are made entirely of polypropylene and are autoclavable to 135°C (275°F)
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 58 mm
Capacity (Metric) 83 mL
Material Polypropylene
Height (Metric) 104 mm
Autoclavable Autoclavable
Description Funnel Buchner PP 55mm
Length (Metric) Stem 45mm
Quantity 1/Pk.; 8/Cs.