Learn More
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
GREENER_CHOICE
Product Code. p-4520587
Click to view available options
Height (Metric):
149 mm
262 mm
Diameter (Metric) Inner:
149 mm
246 mm
Unit Size:
Each
Do not use concentrated acids as desiccants.
Product Code. 10451751
Brand: Thermo Scientific™ 53090250
Only null left
Add to basket
Add to basket
Do not use concentrated acids as desiccants.
View contents easily with these autoclavable polypropylene desiccators, featuring a transparent PC cover and robust body resistant to all solid desiccants.
View contents easily with Thermo Scientific™ Autoclavable Polypropylene Desiccators, featuring a robust body and high resistance to all solid desiccants. These desiccators have a transparent polycarbonate cover. The small size (5315-0150) comes with a 140 mm aluminum plate. The plate for the large size (5309-0250) is sold separately.
  • Clear polycarbonate (PC) cover has recessed rim to keep grease off benchtop when cover is set down
  • No gasket required, only grease is needed
  • Large desiccator (Cat. No. 5310-0250) takes any standard 230mm desiccator plate such as Cat. No. 5312-0230 (sold separately)
  • Autoclavable; repeated autoclaving will reduce the life of the product
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Description Desiccator PP/PC 250mm
Cabinet Style Top Loading
Material Polypropylene, Polycarbonate
Max. Clearance Above Plate 5.6 in.
No. of Shelves 1 (sold separately)
No. per Pack 1
Height (English) 10.32 in.
Height (Metric) 262 mm
Diameter (English) Inner 9.68 in.
Diameter (English) Outer 13 in.
Diameter (Metric) Inner 246 mm
Diameter (Metric) Outer 330 mm