missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAIF1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06197PCP
This item is not returnable.
View return policy
Description
NAIF1 Polyclonal antibody specifically detects NAIF1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| NAIF1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| bA379C10.2, C9orf90RP11-379C10.2, chromosome 9 open reading frame 90, DKFZp762G199, nuclear apoptosis inducing factor 1, nuclear apoptosis-inducing factor 1 | |
| Produced in rabbits immunized with E. coli-derived Human NAIF1 fragment. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication | |
| 203245 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction