missing translation for 'onlineSavingsMsg'
Learn More

NAE1 Monoclonal Antibody (OTI1D12), TrueMAB™, OriGene

Product Code. 17553853
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17553853 30 μL 30µL
17533853 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17553853 Supplier Origene Technologies Supplier No. TA804320S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NAE1 Monoclonal antibody specifically detects NAE1 in Human samples. It is validated for Western Blot

Specifications

Antigen NAE1
Applications Western Blot
Classification Monoclonal
Clone OTI1D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene UFD1
Gene Accession No. Q92890
Gene Alias 59kDa; A-116A10.1; amyloid beta precursor protein binding protein 1, 59kDa; amyloid beta precursor protein-binding protein 1, 59 kDa; amyloid beta precursor protein-binding protein 1, 59kD; amyloid protein-binding protein 1; Appbp1; APP-BP1; HPP1; Nae1; NEDD8 activating enzyme E1 subunit 1; NEDD8-activating enzyme E1 regulatory subunit; NEDD8-activating enzyme E1 subunit; protooncogene protein 1; proto-oncogene protein 1; μga-1
Host Species Mouse
Immunogen Full length human recombinant protein of human UFD1L produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7353
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.