missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAALAD2 Polyclonal Antibody

Description
Immunogen sequence: NDAEILLRYLG GIAPPDKSWK GALNVSYSIG PGFTGSDSFR KVRMHVYNIN Highest antigen sequence identity to the following orthologs - mouse 78%, rat 78%.
Has N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. Also exhibits a dipeptidyl-peptidase IV type activity. Inactivate the peptide neurotransmitter N- acetylaspartylglutamate.
Specifications
Specifications
| Antigen | NAALAD2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NAALAD2 |
| Gene Accession No. | Q9Y3Q0 |
| Gene Alias | GCPIII; Glutamate carboxypeptidase III; GPCIII; NAADALASE2; NAALAD2; NAALADase II; NAALADASE2; N-acetylated alpha-linked acidic dipeptidase 2; N-acetylated alpha-linked acidic dipeptidase II; N-acetylated-alpha-linked acidic dipeptidase 2; N-acetylated-alpha-linked acidic dipeptidase II |
| Gene Symbols | NAALAD2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?