Learn More
Invitrogen™ NA-Star™ Influenza Neuraminidase Inhibitor Resistance Detection Kit
Description
Includes
Substrate, required buffer and reaction accelerator
NA-Star Influenza Neuraminidase Inhibitor Resistance Detection Reagent Set includes everything needed to quantitate neuraminidase activity and neuraminidase inhibitor resistance in avian, equine, human (types A and B), and porcine influenza viruses. The kit's fast and easy protocol and convenient 96-well plate format make it ideal for monitoring influenza virus neuraminidase inhibitor resistance, as well as high-throughput inhibitor compound screening.
- Includes pre-made solutions for all required reagents, enabling you to standardize assays and compare results between labs easily
- Provides up to 50-fold higher sensitivity than assays using fluorescent methylumbelliferone N-acetylneuraminic acid (MUNANA) substrate
- Dynamic range of four orders of magnitude enables you to quantitate virus isolates over a broad range of virus concentration and with varying levels of neuraminidase activity without performing numerous sample dilutions
- Fast and easy protocol enables you to complete assays in less than 1.5 hours, providing rapid assay throughput
Specifications
- Target: Neuraminidase
- Detection Method: Enhanced Chemiluminescence
- For use with Equipment: Luminometer (Microplate)
- Form: Liquid
- High Throughput Compatibility: High Throughput-Compatible
- Readout: End Point
- Substrate Properties: Chemical Substrate
- Substrate Type: Neuraminidase Substrate
Specifications
Specifications
| Content And Storage | This kit contains the enzyme substrate, assay buffer, reaction accelerator, and microplates. Store reagent set components at 2°C to 8°C. |
| Detection Method | Enhanced Chemiluminescence |
| For Use With (Application) | Microbiological Analysis |
| For Use With (Equipment) | Luminometer (Microplate) |
| Product Type | Neuraminidase Assay |
| Form | Liquid |
| Format | 96-well plate |
| Product Line | NA-Star, NovaBright |
| Quantity | 1 kit |
| Substrate | NA-Star™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.