missing translation for 'onlineSavingsMsg'
Learn More

N6,N6-Dimethyladenosine, MedChemExpress

Product Code. 16410067
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16410067 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16410067 Supplier Medchem Express Supplier No. HY101984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N6,N6-Dimethyladenosine is a modified ribonucleoside previously found in rRNA, and also exhibits in mycobacterium bovis Bacille Calmette-Guérin tRNA.

N6,N6-Dimethyladenosine is a modified ribonucleoside previously found in rRNA, and also exhibits in mycobacterium bovis Bacille Calmette-Guérin tRNA.

Specifications

Chemical Name or Material N6,N6-Dimethyladenosine
CAS 2620-62-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H17N5O4
Quantity 50 mg
Solubility Information DMSO : ≥ 125 mg/mL (423.31 mM)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N(C)C)O1)O)O
Molecular Weight (g/mol) 295.29
Formula Weight 295.29
Percent Purity 99.1%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.