missing translation for 'onlineSavingsMsg'
Learn More

N6-Carbobenzyloxy-L-lysine Benzyl Ester Hydrochloride, TRC

Product Code. 30362817
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30362817

Brand: Toronto Research Chemicals TRCC1765251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N6-Carbobenzyloxy-L-lysine Benzyl Ester Hydrochloride
CAS 6366-70-7
Molecular Formula C21H27ClN2O4
Quantity 1 g
Synonym N6-[(Phenylmethoxy)carbonyl]-L-lysine Phenylmethyl Ester Hydrochloride, N6-Carbobenzoxy-L-lysine Benzyl Ester Hydrochloride, NSC 88180
InChI Formula InChI=1S/C21H26N2O4.ClH/c22-19(20(24)26-15-17-9-3-1-4-10-17)13-7-8-14-23-21(25)27-16-18-11-5-2-6-12-18;/h1-6,9-12,19H,7-8,13-16,22H2,(H,23,25);1H/t19-;/m0./s1
SMILES Cl.N[C@@H](CCCCNC(=O)OCc1ccccc1)C(=O)OCc2ccccc2
IUPAC Name benzyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride
Molecular Weight (g/mol) 406.9
Formula Weight 406.1659
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.