missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Carbobenzyloxy-L-lysine Benzyl Ester Hydrochloride, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | N6-Carbobenzyloxy-L-lysine Benzyl Ester Hydrochloride |
| CAS | 6366-70-7 |
| Molecular Formula | C21H27ClN2O4 |
| Quantity | 2 g |
| Synonym | N6-[(Phenylmethoxy)carbonyl]-L-lysine Phenylmethyl Ester Hydrochloride, N6-Carbobenzoxy-L-lysine Benzyl Ester Hydrochloride, NSC 88180 |
| InChI Formula | InChI=1S/C21H26N2O4.ClH/c22-19(20(24)26-15-17-9-3-1-4-10-17)13-7-8-14-23-21(25)27-16-18-11-5-2-6-12-18;/h1-6,9-12,19H,7-8,13-16,22H2,(H,23,25);1H/t19-;/m0./s1 |
| SMILES | Cl.N[C@@H](CCCCNC(=O)OCc1ccccc1)C(=O)OCc2ccccc2 |
| IUPAC Name | benzyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride |
| Molecular Weight (g/mol) | 406.9 |
| Formula Weight | 406.1659 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction