Learn More
N6-(2-Phenylethyl)adenosine, MedChemExpress
Product Code. 16440007
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Product Code. 16440007
Brand: Medchem Express HY101854
MedChemExpress N6-(2-Phenylethyl)adenosine (N6-Phenethyladenosine), an adenosine derivative, is a potent adenosine receptors (AR) agonist with Ki values of 11.8 nM, 30.1 nM, 0.63 nM for rat A1AR, human A1AR and hA3AR, respectively.
N6-(2-Phenylethyl)adenosine (N6-Phenethyladenosine), an adenosine derivative, is a potent adenosine receptors (AR) agonist with Ki values of 11.8 nM, 30.1 nM, 0.63 nM for rat A1AR, human A1AR and hA3AR, respectively.
Specifications
Chemical Name or Material N6-(2-Phenylethyl)adenosine
CAS 20125-39-7
Color White
Purity Grade Notes Research
Molecular Formula C18H21N5O4
Quantity 50 mg
Synonym N6-Phenethyladenosine N6-Phenylethyladenosine
Solubility Information DMSO : 100 mg/mL (269.26 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCCC4=CC=CC=C4)O1)O)O
Molecular Weight (g/mol) 371.39
Formula Weight 371.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation