missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ N4BP2L2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16658224
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16658224 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16658224 Supplier Invitrogen™ Supplier No. PA5101750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total N4BP2L2.

N4BP2L2 (NEDD4-binding protein 2-like 2), also known as PFAAP5 (phosphonoformate immuno-associated protein 5), is a 583 amino acid nuclear protein that potentially is involved in transcriptional regulation. N4BP2L2 is phosphorylated on Ser 199 in response to DNA damage, probably by ATM or ATR. Primarily expressed in bone marrow, N4BP2L2 is dramatically down-regulated after exposure to arsenic compounds, an event which precedes neutropenia. PFAAP5 interacts with both Gfi-1 and Neutrophil Elastase, two proteins that are implicated in neutropenia disorders. Defects in the gene encoding Neutrophil Elastase, ELA2, are the cause of cyclic haematopoiesis, which, with decreased numbers of circulating neutrophils, leads to an increased risk for opportunistic infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP2L2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene N4BP2L2
Gene Accession No. Q8JZS6, Q92802
Gene Alias 2700092H06Rik; 92M18.3; CG005; CG016; N4BP2L2; NEDD4 binding protein 2 like 2; NEDD4 binding protein 2-like 2; NEDD4-binding protein 2-like 2; PFAAP5; phosphonoformate immuno-associated protein 5; Phosphonoformate immuno-associated protein 5 homolog; protein from BRCA2 region; zag1
Gene Symbols N4BP2L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human N4BP2L2(Accession Q92802), corresponding to amino acid residues D358-R408.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10443, 381695
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.