Learn More
N2,N2-Dimethylguanosine, MedChemExpress
Product Code. 16497477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16497477
Brand: Medchem Express HY113137
MedChemExpress N2,N2-Dimethylguanosine is an urinary nucleoside, a primary degradation product of tRNA.
N2,N2-Dimethylguanosine is an urinary nucleoside, a primary degradation product of tRNA.
Specifications
Chemical Name or Material N2,N2-Dimethylguanosine
CAS 2140-67-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H17N5O5
Quantity 5 mg
Solubility Information DMSO : 15 mg/mL (48.19 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=C(N(C)C)NC3=O)O1)O)O
Molecular Weight (g/mol) 311.29
Formula Weight 311.29
Percent Purity 99.31%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death