missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1,N4-Di-p-tolylbenzene-1,4-diamine, TRC
Specifications
Specifications
| Chemical Name or Material | N1,N4-Di-p-tolylbenzene-1,4-diamine |
| CAS | 620-91-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H20 N2 |
| Quantity | 100 mg |
| Synonym | N1,N4-Bis(4-methylphenyl)-1,4-benzenediamine, N,N'-Bis(4-methylphenyl)-p-phenylenediamine, N,N'-Di-p-tolyl-p-phenylenediamine |
| InChI Formula | InChI=1S/C20H20N2/c1-15-3-7-17(8-4-15)21-19-11-13-20(14-12-19)22-18-9-5-16(2)6-10-18/h3-14,21-22H,1-2H3 |
| SMILES | Cc1ccc(Nc2ccc(Nc3ccc(C)cc3)cc2)cc1 |
| IUPAC Name | 1-N,4-N-bis(4-methylphenyl)benzene-1,4-diamine |
| Molecular Weight (g/mol) | 288.386 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?