Learn More
N1-Acetylspermidine Dihydrochloride, TRC
Product Code. 17990355
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990355 5 mg 5mg
17970595 10 mg 10mg
17924555 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17990355 Supplier Toronto Research Chemicals Supplier No. TRCA1878455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34450-16-3
Molecular Formula C9 H21 N3 O . 2 Cl H
Molecular Weight (g/mol) 260.2
Synonym N-[3-[(4-Aminobutyl)amino]propyl]acetamide Hydrochloride (1:2);
IUPAC Name N-[4-(3-aminopropylamino)butyl]acetamide;dihydrochloride
SMILES Cl.Cl.CC(=O)NCCCCNCCCN
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C9H21N3O.2ClH/c1-9(13)12-8-3-2-6-11-7-4-5-10;;/h11H,2-8,10H2,1H3,(H,12,13);2*1H
Formula Weight 259.1218
Percent Purity >95%
Chemical Name or Material N1-Acetylspermidine dihydrochloride
Grade HPLC