missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-N-(4-piperidinyl)propanamide, 97%, admixture with HCl salt

Product Code. 10324122 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 10324122

Brand: Thermo Scientific Acros 363920025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1609-66-1 | C14H20N2O | 232.327 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1609-66-1, 22352-81-4
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C14H20N2O
MDL Number MFCD11106684
Quantity 2.5 g
Synonym norfentanyl, n-phenyl-n-piperidin-4-yl propionamide, n-phenyl-n-piperidin-4-yl-propionamide, 4-n-propionylanilino piperidine, unii-2mk6d8jv6j, n-phenyl-n-4-piperidinyl propanamide, propanamide, n-phenyl-n-4-piperidinyl, n-phenyl-n-4-piperidinylpropanamide, n-phenyl-n-piperidin-4-yl propanamide, n-phenyl-n-4-piperidinylpropionamide
InChI Key PMCBDBWCQQBSRJ-UHFFFAOYSA-N
SMILES CCC(=O)N(C1CCNCC1)C2=CC=CC=C2
IUPAC Name N-phenyl-N-piperidin-4-ylpropanamide
Molecular Weight (g/mol) 232.327
PubChem CID 259381
ChEBI CHEBI:62685
Formula Weight 232.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Phenyl-N-(4-piperidinyl)propanamide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.