Learn More
N-Nitroso Moexipril, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Moexipril |
| CAS | 1026879-47-9 |
| Molecular Formula | C27H33N3O8 |
| Quantity | 25 mg |
| Synonym | (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]nitrosoamino]-1-oxopropyl]-3,4-dihydro-6,7-dimethoxy-3(1H)-isoquinolinecarboxylic acid, (S)-2-(N-((S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl)-N-nitroso-L-alanyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid |
| InChI Formula | InChI=1S/C27H33N3O8/c1-5-38-27(34)21(12-11-18-9-7-6-8-10-18)30(28-35)17(2)25(31)29-16-20-15-24(37-4)23(36-3)14-19(20)13-22(29)26(32)33/h6-10,14-15,17,21-22H,5,11-13,16H2,1-4H3,(H,32,33)/t17-,21-,22-/m0/s1 |
| SMILES | C([C@@H](N([C@@H](CCC1=CC=CC=C1)C(OCC)=O)N=O)C)(=O)N2CC=3C(C[C@H]2C(O)=O)=CC(OC)=C(OC)C3 |
| IUPAC Name | (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]nitrosoamino]-1-oxopropyl]-3,4-dihydro-6,7-dimethoxy-3(1H)-isoquinolinecarboxylic acid |
| Molecular Weight (g/mol) | 527.57 |
| Formula Weight | 527.23 |