missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%

Product Code. 10215671 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
This item is not returnable. View return policy

Product Code. 10215671

Brand: Thermo Scientific Acros 147492500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-18-2 | C10H24N2 | 172.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-18-2
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.31
MDL Number MFCD00008339
InChI Key TXXWBTOATXBWDR-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
PubChem CID 8097
IUPAC Name N,N,N',N'-tetramethylhexane-1,6-diamine
SMILES CN(C)CCCCCCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -46.0°C
Color Colorless to Yellow
Density 0.8060g/mL
Boiling Point 209.0°C to 210.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2N(CH2)6N(CH3)2
Refractive Index 1.4350 to 1.4370
Quantity 250 mL
Beilstein 04, I, 423
Specific Gravity 0.806
Solubility Information Solubility in water: miscible. Other solubilities: soluble in ethanol, acetone, dichloromethane and, toluene
Viscosity 1.72 mPa.s (20°C)
Formula Weight 172.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-1,6-hexanediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.