missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylthioformamide, MP Biomedicals

Product Code. 11428635 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11428635

Brand: MP Biomedicals 0522441410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 758-16-7
Molecular Formula C3H7NS
Quantity 10 g
Synonym n,n-dimethylthioformamide, dimethylthioformamide, methanethioamide, n,n-dimethyl, formamide, n,n-dimethylthio, nn-dimethylthioformamide, methanethioamide,n,n-dimethyl, formamide,n-dimethylthio, dimethylamino methanethial, n,n-dimethyl-thioformamide, methanethioamide, dimethyl
InChI Key SKECXRFZFFAANN-UHFFFAOYSA-N
SMILES CN(C)C=S
IUPAC Name N,N-dimethylmethanethioamide
Molecular Weight (g/mol) 89.156
PubChem CID 69794
Formula Weight 89.16
Chemical Name or Material N, N-Dimethyl Thioformamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.