missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylisobutyramide, 99%

Product Code. p-7035706
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15420238

Brand: Thermo Scientific Alfa Aesar H53430.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21678-37-5 | C6H13NO | 115.176 g/mol

1- chloro-N,N-2-trimethylpropenylamine readily prepared' from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1- chloro-N,N-2-trimethylpropenylamine readily prepared′ from N,N-dimethylisobutyramide. Isoxazoles are prepared by deprotonation of N,N-dimethylisobutyramide.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 21678-37-5
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00075378
InChI Key GXMIHVHJTLPVKL-UHFFFAOYSA-N
Synonym n,n-dimethylisobutyramide, n,n,2-trimethylpropionamide, n,n-dimethyl-isobutyramide, acmc-20ak9x, n,n-dimethylisobutylamide, isobutyric acid dimethylamide, propanamide, n,n,2-trimethyl, propanamide,n,n,2-trimethyl, n,n-dimethyl2-methylpropionamide
PubChem CID 243415
IUPAC Name N,N,2-trimethylpropanamide
SMILES CC(C)C(=O)N(C)C

Specifica

Density 0.892
Boiling Point 175°C to 176°C (44 mmHg)
Flash Point 154°C (309°F)
Linear Formula (CH3)2CHCON(CH3)2
Refractive Index 1.439
Quantity 5 g
Beilstein 1742178
Solubility Information Soluble in water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material N,N-Dimethylisobutyramide
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato