missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldecylamine N-Oxide, TRC

Product Code. 30378401
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378401 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378401 Supplier Toronto Research Chemicals Supplier No. TRCD47249810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethyldecylamine N-Oxide
CAS 2605-79-0
Molecular Formula C12H27NO
Quantity 10 g
Synonym Ammonyx C 10, Ammonyx DO, Ammonyx DO-C 10, Amphitol 10N, Aromox DM 10DW, Barlox 10S, Decyl-N,N-dimethylamine Oxide, Decyldimethylamine N-Oxide, Decyldimethylamine Oxide, Dimethyl decylamine Oxide, Euroxide DO 40, Genaminox DC 40, Genaminox K 10, Mackamine 065, N,N-Dimethyl-1-decanamine Oxide, N,N-Dimethyl-N-decylamine N-Oxide, N,N-Dimethyl-n-decylamine Oxide, N-Decyl-N,N-dimethylamine Oxide, Tegotens DO
InChI Formula InChI=1S/C12H27NO/c1-4-5-6-7-8-9-10-11-12-13(2,3)14/h4-12H2,1-3H3
SMILES CCCCCCCCCC[N+](C)(C)[O-]
IUPAC Name N,N-dimethyldecan-1-amine oxide
Molecular Weight (g/mol) 201.35
Formula Weight 201.2093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.