missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide, 99%

Product Code. p-7044093
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11301617

Brand: Thermo Scientific Alfa Aesar L04503.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-84-5 | C5H11NO | 101.15 g/mol

N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylformamide is used in the synthesis of metal-organic frameworks and Diethyltrifluoromethylamine. It is also used as solvent in the preparation of porous cubic-shaped zinc oxide particles. It is involved in the preparation of quinazoline-2,4(1H,3H)-dione by reacting with o-aminonitrile.

Solubility
Miscible with acetone, benzene, alcohol and ether.  

Notes
Incompatible with strong oxidizing agents. Avoid heat, flames and spark.                      
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-84-5
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003287
InChI Key SUAKHGWARZSWIH-UHFFFAOYSA-N
Synonym diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
PubChem CID 12051
IUPAC Name N,N-diethylformamide
SMILES CCN(CC)C=O

Specifications

Density 0.91
Boiling Point 178°C
Flash Point 65°C (149°F)
Odor Amine-like
Refractive Index 1.434
Quantity 100 g
Beilstein 1209392
Solubility Information Miscible with acetone,benzene,alcohol and ether.
Formula Weight 101.15
Percent Purity 99%
Chemical Name or Material N,N-Diethylformamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylformamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Diethylformamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.