Product Code. 10236523
Shop All Thermo Scientific Chemicals Products
Click to view available options
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
10236523
2.5 L
Glass bottle
2.5 L
10050880
1 L
Glass bottle
1 L
13458308
10 L
Plastic drum
10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
10236523
Supplier
Thermo Scientific Acros
Supplier No.
114330025
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
100-37-8
MDL Number
MFCD00002850
InChI Key
BFSVOASYOCHEOV-UHFFFAOYSA-N
Synonym
ethanol, 2-diethylamino, 2-hydroxyethyl diethylamine, n,n-diethylethanolamine, 2-diethylamino ethanol, deae, diethylethanolamine, diethylamino ethanol, n,n-diethyl-2-aminoethanol, diethylaminoethanol, diethyl 2-hydroxyethyl amine
PubChem CID
7497
ChEBI
CHEBI:52153
SMILES
CCN(CC)CCO
Specifications
CAS Min %
98.5
CAS Max %
100.0
Melting Point
-70.0°C
Color
White to Yellow
Density
0.8800 g/mL
Boiling Point
161.0°C
Flash Point
51.5°C
Infrared Spectrum
Authentic
Assay Percent Range
98.5% min. (GC)
Packaging
Glass bottle
Molecular Formula
C6H15NO
Linear Formula
HOCH2 CH2 N(C2 H5 )2
Refractive Index
1.4400 to 1.4420
Quantity
2.5 L
Beilstein
04,282
Merck Index
15,3123
Specific Gravity
0.88
Solubility Information
Solubility aq. soln.: soluble. Other solubilities: soluble in alcohol, ether and benzene
IUPAC Name
2-(diethylamino)ethan-1-ol
Molecular Weight (g/mol)
117.19
Formula Weight
117.19
Percent Purity
99%
Physical Form
Crystalline Powder
Chemical Name or Material
N, N-Diethylethanolamine, 99%
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?