missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals

Product Code. p-109162 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
missing translation for 'unitSize'
1 L
10 L
2.5 L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10050880

missing translation for 'mfr': Thermo Scientific Acros 114330010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-37-8
MDL Number MFCD00002850
InChI Key BFSVOASYOCHEOV-UHFFFAOYSA-N
Synonym 2-diethylamino ethanol, n,n-diethylethanolamine, diethylaminoethanol, deae, diethylethanolamine, diethylamino ethanol, ethanol, 2-diethylamino, n,n-diethyl-2-aminoethanol, 2-hydroxyethyl diethylamine, diethyl 2-hydroxyethyl amine
PubChem CID 7497
ChEBI CHEBI:52153
SMILES CCN(CC)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Color White to Yellow
Density 0.8800g/mL
Boiling Point 161.0°C
Flash Point 51.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H15NO
Linear Formula HOCH2CH2N(C2H5)2
Refractive Index 1.4400 to 1.4420
Quantity 1 L
Beilstein 04,282
Merck Index 15,3123
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and benzene
IUPAC Name 2-(diethylamino)ethanol
Molecular Weight (g/mol) 117.19
Formula Weight 117.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Diethylethanolamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylethanolamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.