Learn More
N,N-Di-n-octylaniline, 98%
Product Code. 11352539
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352539 1 g 1g
11362539 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11352539 Supplier Thermo Scientific Alfa Aesar Supplier No. H26644.03
Request Bulk or Custom Format
CAS: 3007-75-8 | C22H39N | 317.56 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3007-75-8
Molecular Formula C22H39N
Molecular Weight (g/mol) 317.56
MDL Number MFCD00155305
InChI Key PTRSTXBRQVXIEW-UHFFFAOYSA-N
Synonym n,n-di-n-octylaniline, benzenamine, n,n-dioctyl, dioctylphenylamine, benzenamine,n,n-dioctyl
PubChem CID 137791
IUPAC Name N,N-dioctylaniline
SMILES CCCCCCCCN(CCCCCCCC)C1=CC=CC=C1
Specifications
Refractive Index 1.5495
Quantity 1 g
Formula Weight 317.56
Percent Purity 98%
Chemical Name or Material N,N-Di-n-octylaniline