Learn More
N,N-Bis(2-chloroethyl)aniline, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Bis(2-chloroethyl)aniline |
| CAS | 553-27-5 |
| Molecular Formula | C10H13NCl2 |
| Quantity | 50 mg |
| Synonym | N,N-Bis(2-chloroethyl)benzenamine, Aniline mustard, Lymphochin, Lymphocin, Lymphoquin, N,N-Bis(2'-chloroethyl)aniline, N,N-Di(2-chloroethyl)aniline, NCS-18429, NSC 18429, beta,beta'-Dichlorodiethylaniline |
| InChI Formula | InChI=1S/C10H13Cl2N/c11-6-8-13(9-7-12)10-4-2-1-3-5-10/h1-5H,6-9H2 |
| SMILES | ClCCN(CCCl)c1ccccc1 |
| IUPAC Name | N,N-bis(2-chloroethyl)aniline |
| Molecular Weight (g/mol) | 218.12 |
| Formula Weight | 217.0425 |