missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzenesulfonamide, 97%

Product Code. p-7037089
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15471978

Brand: Thermo Scientific Alfa Aesar H55666.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5183-78-8 | C7H9NO2S | 171.21 g/mol

N-Methylbenzenesulfonamide, is used as an intermediate in industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylbenzenesulfonamide, is used as an intermediate in industry.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5183-78-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025023
InChI Key SVDVKEBISAOWJT-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-methyl, n-methylbenzenesulphonamide, n-phenylsulfonyl methanamine, methyl phenylsulfonyl amine, n-methylphenylsulfonamide, acmc-1avn9, n-methylbenzene sulfonamide, n-methyl benzenesulphonamide, benzenesulfonamide,n-methyl, benzenesulfonyl-n-methylamide
PubChem CID 78858
IUPAC Name N-methylbenzenesulfonamide
SMILES CNS(=O)(=O)C1=CC=CC=C1

Specifications

Quantity 250 mg
Solubility Information It is soluble in water.
Formula Weight 171.21
Percent Purity 97%
Chemical Name or Material N-Methylbenzenesulfonamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.