missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzamide, 99%

Product Code. 11429206
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11429206

Brand: Thermo Scientific Alfa Aesar A18057.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 613-93-4 | C8H9NO | 135.17 g/mol

It acts as a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as a potent PDE10A (phosphodiesterase with a remarkable localization as the protein is abundant only in brain tissue) inhibitor. It is also employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-93-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00011642
InChI Key NCCHARWOCKOHIH-UHFFFAOYSA-N
Synonym benzamide, n-methyl, n-methylbenzenamide, n-methyl-benzamide, n-methylbenzenecarboxamide, unii-k3ed781e08, ccris 4670, n-methyl benzamide, n-methylbenzimidic acid, acmc-1bb7i, dsstox_cid_5570
PubChem CID 11954
IUPAC Name N-methylbenzamide
SMILES CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 76°C to 81°C
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material N-Methylbenzamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.