missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl Serotonin Oxalate Salt, TRC
Specifications
Specifications
| CAS | 1975-81-1 |
| Molecular Formula | C11 H14 N2 O . C2 H2 O4 |
| Quantity | 10 mg |
| Synonym | 1H-Indol-5-ol, 3-[2-(methylamino)ethyl]-, ethanedioate (1:1), 1H-Indol-5-ol, 3-[2-(methylamino)ethyl]-, ethanedioate (1:1) (salt) (9CI), Indol-5-ol, 3-[2-(methylamino)ethyl]-, oxalate (1:1) (salt) (8CI), N-Methyl-5-hydroxytryptamine hydrogen oxalate, N-Methylserotonin oxalate, NSC 91541 |
| InChI Formula | InChI=1S/C11H14N2O.C2H2O4/c1-12-5-4-8-7-13-11-3-2-9(14)6-10(8)11;3-1(4)2(5)6/h2-3,6-7,12-14H,4-5H2,1H3;(H,3,4)(H,5,6) |
| SMILES | CNCCc1c[nH]c2ccc(O)cc12.OC(=O)C(=O)O |
| IUPAC Name | 3-[2-(methylamino)ethyl]-1H-indol-5-ol;oxalic acid |
| Molecular Weight (g/mol) | 280.28 |
| Formula Weight | 280.1059 |
| Chemical Name or Material | N-Methyl Serotonin Oxalate salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?