missing translation for 'onlineSavingsMsg'
Learn More

N-L-α-Glutamyl-L-glutamic Acid, TRC

Product Code. 30341662
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30341662 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341662 Supplier Toronto Research Chemicals Supplier No. TRCA7902735MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-2-((S)-2-Amino-4-carboxybutanamido)pentanedioic Acid
CAS 3929-61-1
Molecular Formula C10H16N2O7
Quantity 5 mg
Synonym (S)-2-((S)-2-Amino-4-carboxybutanamido)pentanedioic Acid, L-N-L-alpha-Glutamyl-glutamic Acid (8CI), N-L-alpha-Glutamyl-L-glutamic Acid, L-alpha-Glutamyl-L-glutamic Acid, 107: PN: US20070066537PAGE: 17 claimed protein, 2: PN: WO2008083678 PAGE: 33 claimed protein, 84: PN: EP2161028 PAGE: 10 claimed protein, 90: PN: WO2004025259PAGE: 57 claimed protein, L-Glutamyl-L-glutamic Acid, NSC 335983, alpha-Glutamylglutamic Acid
InChI Formula InChI=1S/C10H16N2O7/c11-5(1-3-7(13)14)9(17)12-6(10(18)19)2-4-8(15)16/h5-6H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)/t5-,6-/m0/s1
SMILES N[C@@H](CCC(=O)O)C(=O)N[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2S)-2-[[(2S)-2-amino-4-carboxybutanoyl]amino]pentanedioic acid
Molecular Weight (g/mol) 276.24
Formula Weight 276.0958
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.