Learn More
n-Hexylbenzene-2,3,4,5,6-d5, CDN
Product Code. 30309221
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309221 0.5 g 0.5g
30325732 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309221 Supplier CDN Supplier No. CDND67330.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material n-Hexylbenzene-2,3,4,5,6-D5
CAS 1219799-07-1
Molecular Formula C12 D5 H13
Quantity 0.5 g
Synonym Benzene-1,2,3,4,5-d5, 6-hexyl- (ACI), 6-Hexylbenzene-1,2,3,4,5-d5 (ACI), n-Hexylbenzene-2,3,4,5,6-D5
InChI Formula InChI=1 S/C12H18/c1-2-3-4-6-9-12-10-7-5-8-11-12/h5,7-8,10-11 H,2-4,6,9H2,1H3/i5D,7 D,8 D,10 D,11 D
SMILES [2 H]c1c([2 H])c([2 H])c(CCCCCC)c([2 H])c1[2 H]
IUPAC Name 1,2,3,4,5-pentadeuterio-6-hexylbenzene
Molecular Weight (g/mol) 167.302
Formula Weight 167.172 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity