Learn More
N-desethyl-N-(tert-butyl)-Terbuthylazine, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(tert-butylamino)chloro-s-triazine |
| CAS | 39605-42-0 |
| Molecular Formula | C11H20ClN5 |
| Quantity | 1 g |
| Synonym | Bis(tert-butylamino)chloro-s-triazine, 6-Chloro-N2,N4-bis(1,1-dimethylethyl)-1,3,5-triazine-2,4-diamine, 2,4-Bis(tert-butylamino)-6-chloro-1,3,5-triazine, 6-Chloro-N,N'-bis(1,1-dimethylethyl)- 1,3,5-Triazine-2,4-diamine |
| InChI Formula | InChI=1S/C11H20ClN5/c1-10(2,3)16-8-13-7(12)14-9(15-8)17-11(4,5)6/h1-6H3,(H2,13,14,15,16,17) |
| SMILES | ClC1=NC(NC(C)(C)C)=NC(NC(C)(C)C)=N1 |
| IUPAC Name | N2,N4-di-tert-butyl-6-chloro-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 257.76 |
| Formula Weight | 257.14 |