Learn More
N-Boc-N-deshydroxyethyl Dasatinib, TRC
Specifications
Specifications
| Chemical Name or Material | N-Boc-N-deshydroxyethyl Dasatinib |
| CAS | 1159977-13-5 |
| Molecular Formula | C25H30ClN7O3S |
| Quantity | 25 mg |
| Synonym | 1-Piperazinecarboxylic acid, 4-[6-[[5-[[(2-chloro-6-methylphenyl)amino]carbonyl]-2-thiazolyl]amino]-2-methyl-4-pyrimidinyl]-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 4-[6-[[5-[[(2-chloro-6-methylphenyl)amino]carbonyl]-2-thiazolyl]amino]-2-methyl-4-pyrimidinyl]-1-piperazinecarboxylate (ACI), 1-Piperazinecarboxylic acid, 4-[6-[[5-[[(2-chloro-6-methylphenyl)amino]carbonyl]-2-thiazolyl]amino]-2-methyl-4-pyrimidinyl]-, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 4-[6-[[5-[[(2-chloro-6-methylphenyl)amino]carbonyl]-2-thiazolyl]amino]-2-methyl-4-pyrimidinyl]-1-piperazinecarboxylate (ACI) |
| InChI Formula | InChI=1S/C25H30ClN7O3S/c1-15-7-6-8-17(26)21(15)31-22(34)18-14-27-23(37-18)30-19-13-20(29-16(2)28-19)32-9-11-33(12-10-32)24(35)36-25(3,4)5/h6-8,13-14H,9-12H2,1-5H3,(H,31,34)(H,27,28,29,30) |
| SMILES | Cc1cccc(Cl)c1NC(=O)c2cnc(Nc3cc(nc(C)n3)N4CCN(CC4)C(=O)OC(C)(C)C)s2 |
| IUPAC Name | tert-butyl 4-[6-[[5-[(2-chloro-6-methylphenyl)carbamoyl]-1,3-thiazol-2-yl]amino]-2-methylpyrimidin-4-yl]piperazine-1-carboxylate |
| Molecular Weight (g/mol) | 544.07 |
| Formula Weight | 543.1819 |