missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-L-isoleucine hemihydrate, 98+%

Product Code. 15477727
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15477727

Brand: Alfa Aesar™ H37050.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 204138-23-8
Molecular Formula C22H44N2O9
Molecular Weight (g/mol) 480.599
MDL Number MFCD00672705
InChI Key NYGJSARLXBZKTA-AQHOMMPKSA-N
Synonym boc-l-isoleucine hemihydrate, boc-ile-oh 1/2h2o, boc-isoleucine hemihydrate, boc-l-isoleucine 1/2h2o, n-boc-l-isoleucine hemihydrate, n-boc-l-isoleucine hemihydrate, 98+%, n-t-butyloxycarbonyl-l-isoleucine hemihydrate, n-tert-butoxycarbonyl-l-isoleucine hemihydrate, n-alpha-t-butyloxycarbonyl-l-isoleucine semihydrate, l-isoleucine, n-1,1-dimethylethoxy carbonyl-, hydrate 2:1
PubChem CID 45072434
IUPAC Name (2S,3S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
SMILES CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.CCC(C)C(C(=O)O)NC(=O)OC(C)(C)C.O

Specifications

Melting Point 67°C to 70°C
Quantity 5 g
Formula Weight 240.07 (231.06 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material N-Boc-L-isoleucine hemihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.