missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-beta-prolinol, 97+%, Thermo Scientific™

Product Code. 15474178
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15474178

Brand: Thermo Scientific Alfa Aesar H57753.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199174-24-8
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD03094726
InChI Key HKIGXXRMJFUUKV-QMMMGPOBSA-N
Synonym s-tert-butyl 3-hydroxymethyl pyrrolidine-1-carboxylate, s-1-boc-3-hydroxymethyl pyrrolidine, s-1-boc-3-hydroxymethyl-pyrrolidine, s-3-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3s-3-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-s-3-hydroxymethyl pyrrolidine, s-1-boc-3-pyrrolidinemethanol, s-n-boc-pyrrolidine-3-methanol, 1-boc-3 s-hydroxymethyl pyrrolidine
PubChem CID 1514341
IUPAC Name tert-butyl (3S)-3-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)CO

Specifications

Density 1.084
Boiling Point 285°C to 289°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 201.27
Percent Purity ≥97%
Chemical Name or Material N-Boc-L-beta-prolinol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.