Learn More
N-Boc-4-bromo-3-fluoroaniline, 96%, Thermo Scientific Chemicals
Product Code. 15485698
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485698 250 mg 250mg
15475698 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15485698 Supplier Thermo Scientific Alfa Aesar Supplier No. H59398.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 868735-43-7
Molecular Formula C11H13BrFNO2
Molecular Weight (g/mol) 290.132
MDL Number MFCD11976284
InChI Key FQLYUJLJZFATNI-UHFFFAOYSA-N
Synonym tert-butyl 4-bromo-3-fluorophenyl carbamate, tert-butyl n-4-bromo-3-fluorophenyl carbamate, tert-butyl 4-bromo-3-fluorophenylcarbamate, 4-bromo-3-fluoro-phenyl-carbamic acid tert-butyl ester, n-boc-4-bromo-3-fluoroaniline
PubChem CID 59852943
IUPAC Name tert-butyl N-(4-bromo-3-fluorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC(=C(C=C1)Br)F
Specifications
Melting Point 68°C to 70°C
Quantity 250 mg
UN Number UN2811
Beilstein 14460051
Formula Weight 290.13
Percent Purity 96%
Chemical Name or Material N-Boc-4-bromo-3-fluoroaniline