Learn More
N-Boc-3-aminopropanol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Boc-3-aminopropanol |
| CAS | 58885-58-8 |
| Molecular Formula | C8H17NO3 |
| Quantity | 1 g |
| Synonym | N-(3-Hydroxypropyl)-carbamic acid 1,1-Dimethylethylester, (3-Hydroxypropyl)carbamic Acid 1,1-Dimethylethylester, (3-Hydroxypropyl)carbamic Acid tert-Butylester, 1,1-Dimethylethyl (3-Hydroxypropyl)carbamate, 3-(Boc-amino)-1-propanol, 3-(N-Boc-amino)-1-propanol, 3-(N-tert-Butoxycarbonylamino)-1-propanol, 3-(N-tert-Butoxycarbonylamino)propanol, 3-(tert-Butoxycarbonylamino)-1-propanol, 3-(tert-Butoxycarbonylamino)propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butylester, N-(3-Hydroxypropyl)carbamic Acid tert-Butylester, N-(tert-Butoxycarbonyl)-3-amino-1-propanol, tert-Butyl(3-hydroxypropyl)carbamate, tert-Butyl-N-(3-hydroxypropyl)carbamate |
| InChI Formula | InChI=1S/C8H17NO3/c1-8(2,3)12-7(11)9-5-4-6-10/h10H,4-6H2,1-3H3,(H,9,11) |
| SMILES | CC(C)(C)OC(=O)NCCCO |
| IUPAC Name | tert-butyl N-(3-hydroxypropyl)carbamate |
| Molecular Weight (g/mol) | 175.23 |
| Formula Weight | 175.1208 |