missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,3-diaminopropane, 95%

Product Code. 11439460
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439460 1 g 1g
11449460 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439460 Supplier Thermo Scientific Alfa Aesar Supplier No. H50304.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75178-96-0 | C8H19N2O2 | 175.25 g/mol

N-Boc-1,3-diaminopropane is used in the preparation of spermidine analogues as well as in the preparation of pharmacologically active compounds. It is also used in suzuki reaction. Further, it is used in the preparation of [3-(3-cyano-propylamino)-propyl]-carbamic acid tert-butyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,3-diaminopropane is used in the preparation of spermidine analogues as well as in the preparation of pharmacologically active compounds. It is also used in suzuki reaction. Further, it is used in the preparation of [3-(3-cyano-propylamino)-propyl]-carbamic acid tert-butyl ester.

Solubility
Miscible with methanol.

Notes
Hygroscopic. Air sensitive. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75178-96-0
Molecular Formula C8H19N2O2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00210021
InChI Key POHWAQLZBIMPRN-UHFFFAOYSA-O
Synonym n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
PubChem CID 2735700
SMILES CC(C)(C)OC(=O)NCCC[NH3+]

Specifications

Melting Point ∼22°C
Linear Formula (CH3)3COCONHCH2CH2CH2NH2
UN Number UN3259
Solubility Information Miscible with methanol.
IUPAC Name tert-butyl N-(3-azaniumylpropyl)carbamate
Quantity 1 g
Formula Weight 174.24
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material N-Boc-1,3-diaminopropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.