missing translation for 'onlineSavingsMsg'
Learn More

N-Bis(2-hydroxypropyl)nitrosamine, MedChemExpress

Product Code. 16473126
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16473126

Brand: Medchem Express HY112085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Bis(2-hydroxypropyl)nitrosamine is an agent with carcinogenic activity.

N-Bis(2-hydroxypropyl)nitrosamine is an agent with carcinogenic activity.

Specifications

Chemical Name or Material N-Bis(2-hydroxypropyl)nitrosamine
Physical Form Oil
CAS 53609-64-6
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C6H14N2O3
For Use With (Application) Cancer-programmed cell death
Synonym DHPN Di(2-hydroxypropyl)nitrosamine Diisopropanolnitrosamine
Solubility Information DMSO : ≥ 125 mg/mL (770.70 mM)
SMILES O=NN(CC(O)C)CC(O)C
Molecular Weight (g/mol) 162.19
Formula Weight 162.19
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.